2,5-Di(2-thienyl)thieno[3,2-b]thiophene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Di(thiophen-2-yl)thieno[3,2-b]thiophene

A703648-100MG A703648-1G A703648-250MG A703648-5G
77018-750EA 39.26 USD
77018-750 77018-752 77018-754 77018-756
2,5-Di(2-thienyl)thieno[3,2-b]thiophene 98%
2,5-Di(2-thienyl)thieno[3,2-b]thiophene

Precaución: for Research Use Only

Formula: C₁₄H₈S₄
MW: 304.46 g/mol
MDL Number: MFCD11114556
CAS Number: 21210-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR