1-Methyl-3-(4-sulfobutyl)imidazolium Hydrogen sulfate 99%

Supplier: Ambeed
Danger

Synonyms: 1-butylsulphonic-3-methylimidazolium hydrogensulphate

A703493-25G A703493-5G
76992-698EA 46 USD
76992-698 76992-700
1-Methyl-3-(4-sulfobutyl)imidazolium Hydrogen sulfate 99%
1-Methyl-3-(4-sulfobutyl)imidazolium Hydrogen sulfate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₇S₂
MDL Number: MFCD12761445
CAS Number: 827320-59-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR