N-(3-Bromo-4-fluorophenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A703145-1G A703145-10G
77018-748EA 34 USD
77018-748 77891-383
N-(3-Bromo-4-fluorophenyl)methanesulfonamide 98%
N-(3-Bromo-4-fluorophenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₇BrFNO₂S
MDL Number: MFCD18071053
CAS Number: 1256633-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR