Ethyl 4-(trifluoromethyl)nicotinate 95%

Supplier: Ambeed
Warning

A703030-1G A703030-250MG
77035-790EA 164 USD
77035-790 77035-792
Ethyl 4-(trifluoromethyl)nicotinate 95%
Ethyl 4-(trifluoromethyl)nicotinate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MDL Number: MFCD11977214
CAS Number: 1214332-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR