8-Methylquinolin-5-amine 95%

Supplier: Ambeed
Warning

A702852-1G A702852-250MG
77005-986EA 115.64 USD
77005-986 77005-988
8-Methylquinolin-5-amine 95%
8-Methylquinolin-5-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.19 g/mol
MDL Number: MFCD03724045
CAS Number: 50358-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR