1,4-Dimethyl-2-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1,4-dimethylbenzene, 2-Fluoro-p-xylene

A702022-100G A702022-1G A702022-25G A702022-5G
77035-778EA 166 USD
77035-778 77035-780 77035-782 77035-784
1,4-Dimethyl-2-fluorobenzene 98%
1,4-Dimethyl-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₉F
MW: 124.155 g/mol
MDL Number: MFCD03412243
CAS Number: 696-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR