(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride, Sapropterin 2HCl, Biopten, Sapropterin dihydrochloride, (R)-2-Amino-6-((1R,2S)-1,2-dihydroxypropyl)-5,6,7,8-tetrahydropteridin-4(1H)-one dihydrochloride

A701042-10MG A701042-1MG A701042-50MG A701042-100MG A701042-1G A701042-250MG
77005-974EA 59 USD
77005-974 77005-976 77005-978 77673-920 77882-561 77882-562
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride 95%
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅N₅O₃·2HCl
MDL Number: MFCD00891665
CAS Number: 69056-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR