Z-HomoPhe-OH 97%

Supplier: Ambeed
Warning

A700842-1G A700842-5G
77035-776EA 6 USD
77035-776 77873-105
Z-HomoPhe-OH 97%
Z-HomoPhe-OH

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₄
MDL Number: MFCD00038796
CAS Number: 127862-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR