Methyl-4-amino-3-bromo-5-fluorobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-3-bromo-5-fluorobenzoate

A700521-250MG A700521-1G
77018-732EA 18 USD
77018-732 77891-375
Methyl-4-amino-3-bromo-5-fluorobenzoate 95%
Methyl-4-amino-3-bromo-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
MW: 248.06 g/mol
MDL Number: MFCD11846084
CAS Number: 1123171-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR