5-Amino-2-cyano-3-methylpyridine 95%
Supplier: Ambeed
|
Warning
|
A700272-1G
A700272-250MG
A700272-5G
77018-726EA
123
USD
77018-726
77018-728
77018-730
5-Amino-2-cyano-3-methylpyridine 95%
5-Amino-2-cyano-3-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₇N₃ |
MDL Number:
MFCD07375122 CAS Number: 252056-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...