Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 98%

Supplier: Ambeed
Warning

Synonyms: 1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane, Trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane, Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane

A699819-5G A699819-100G
77018-720EA 35 USD
77018-720 77614-170
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 98%
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane

Precaución: for Research Use Only

Formula: C₉H₁₃F₉O₃Si
MW: 368.27 g/mol
MDL Number: MFCD08275518
CAS Number: 85877-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR