4-Bromo-1H-indazol-3(2H)-one 98%

Supplier: Ambeed
Warning

A699516-100MG A699516-1G A699516-250MG
77018-714EA 41.38 USD
77018-714 77018-716 77018-718
4-Bromo-1H-indazol-3(2H)-one 98%
4-Bromo-1H-indazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.03 g/mol
MDL Number: MFCD13176950
CAS Number: 864845-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR