5-Bromo-6-chloro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Danger

A698638-100MG A698638-1G A698638-250MG A698638-5G
76992-686EA 38.2 USD
76992-686 76992-688 76992-690 77614-162
5-Bromo-6-chloro-1H-pyrrolo[2,3-b]pyridine 97%
5-Bromo-6-chloro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.47 g/mol
MDL Number: MFCD12963482
CAS Number: 1190321-59-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR