Ethyl 1-Boc-3-ethylpiperidine-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A697158-1G
A697158-250MG
A697158-5G
77018-706EA
68.96
USD
77018-706
77018-708
77018-710
Ethyl 1-Boc-3-ethylpiperidine-3-carboxylate 95%
Ethyl 1-Boc-3-ethylpiperidine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₂₇NO₄ MW: 285.37 g/mol |
MDL Number:
MFCD21641311 CAS Number: 1178882-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...