1-(4-Fluorophenyl)cyclopropanecarbonitrile 98%
Supplier: Ambeed
|
Danger
|
A696648-10G
A696648-1G
A696648-250MG
A696648-25G
A696648-5G
76992-676EA
57
USD
76992-676
76992-678
76992-680
76992-682
76992-684
1-(4-Fluorophenyl)cyclopropanecarbonitrile 98%
1-(4-Fluorophenyl)cyclopropanecarbonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₈FN |
MDL Number:
MFCD01314307 CAS Number: 97009-67-1 UN: 3276 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...