tert-Butyl 3-iodo-1H-pyrazolo[3,4-b]pyridine-1-carboxylate 95+%

Supplier: Ambeed
Warning

A696037-100MG A696037-1G A696037-250MG
77018-700EA 15.92 USD
77018-700 77018-702 77018-704
tert-Butyl 3-iodo-1H-pyrazolo[3,4-b]pyridine-1-carboxylate 95+%
tert-Butyl 3-iodo-1H-pyrazolo[3,4-b]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂IN₃O₂
MW: 345.13 g/mol
MDL Number: MFCD13183727
CAS Number: 920036-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR