(3-(Ethylsulfonamido)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A696030-1G A696030-250MG A696030-5G
76992-670EA 157.02 USD
76992-670 76992-672 76992-674
(3-(Ethylsulfonamido)phenyl)boronic acid 98%
(3-(Ethylsulfonamido)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
MDL Number: MFCD10699609
CAS Number: 710348-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR