3-Bromo-6-methyl-1H-indazol-5-amine 95%

Supplier: Ambeed
Warning

A695980-100MG A695980-1G
77018-690EA 124.13 USD
77018-690 77018-692
3-Bromo-6-methyl-1H-indazol-5-amine 95%
3-Bromo-6-methyl-1H-indazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₈BrN₃
MW: 226.07 g/mol
MDL Number: MFCD09026984
CAS Number: 1000343-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR