(1R,3R,5R)-2-[(2-Methylpropan-2-yl)oxycarbonyl]-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (1R,3R,5R)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

A693924-250MG A693924-1G A693924-100MG
77018-682EA 79 USD
77018-682 77684-601 77891-362
(1R,3R,5R)-2-[(2-Methylpropan-2-yl)oxycarbonyl]-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 98%, Ambeed.Inc
(1R,3R,5R)-2-[(2-Methylpropan-2-yl)oxycarbonyl]-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

Precaución: For research use only.

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD16621713
CAS Number: 1148048-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR