Rhodium(II) 2,2,2-triphenylacetate 97%

Supplier: Ambeed
Warning

A693247-100MG A693247-250MG
77035-746EA 72.15 USD
77035-746 77035-748
Rhodium(II) 2,2,2-triphenylacetate 97%
Rhodium(II) 2,2,2-triphenylacetate

Precaución: for Research Use Only

Formula: C₄₀H₃₀O₄Rh
MW: 677.56 g/mol
MDL Number: MFCD00191694
CAS Number: 68803-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR