4-Chloroquinolin-6-amine 97%

Supplier: Ambeed
Warning

A693061-100MG A693061-1G A693061-250MG
77035-740EA 114.58 USD
77035-740 77035-742 77035-744
4-Chloroquinolin-6-amine 97%
4-Chloroquinolin-6-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MW: 178.618 g/mol
MDL Number: MFCD14702408
CAS Number: 1085192-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR