4-Methyl-1H-indazol-7-amine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Amino-4-methyl-1H-indazole
A691991-100MG
A691991-1G
A691991-250MG
77005-936EA
28.65
USD
77005-936
77005-938
77005-940
4-Methyl-1H-indazol-7-amine 95%
4-Methyl-1H-indazol-7-amine
Precaución: for Research Use Only
Formula:
C₈H₉N₃ MW: 147.18 g/mol |
MDL Number:
MFCD09607935 CAS Number: 952183-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...