Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 98+%

Supplier: Ambeed
Warning

A691636-100MG A691636-1G A691636-250MG
77035-734EA 71.09 USD
77035-734 77035-736 77035-738
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate 98+%
Methyl 4-ethyl-2-methyl-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.193 g/mol
MDL Number: MFCD12756158
CAS Number: 1245644-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR