Methyl 3-bromo-4,5-difluorobenzoate 97%

Supplier: Ambeed
Warning

A690737-1G A690737-5G
77018-670EA 26.53 USD
77018-670 77018-672
Methyl 3-bromo-4,5-difluorobenzoate 97%
Methyl 3-bromo-4,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
MW: 251.025 g/mol
MDL Number: MFCD20483155
CAS Number: 1244642-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR