Hex-5-yn-1-amine 98%

Supplier: Ambeed
Danger

A690012-1G A690012-250MG A690012-100MG A690012-5G
76992-650EA 126 USD
76992-650 76992-652 77614-128 77891-358
Hex-5-yn-1-amine 98%
Hex-5-yn-1-amine

Precaución: for Research Use Only

Formula: C₆H₁₁N
MDL Number: MFCD09923614
CAS Number: 15252-45-6
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR