5-Bromo-6-(trifluoromethyl)-4-pyrimidinol 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-(trifluoromethyl)pyrimidin-4-ol, 5-Bromo-4-hydroxy-6-(trifluoromethyl)pyrimidine

A689401-1G A689401-250MG A689401-5G
77005-922EA 43.5 USD
77005-922 77005-924 77005-926
5-Bromo-6-(trifluoromethyl)-4-pyrimidinol 97%
5-Bromo-6-(trifluoromethyl)-4-pyrimidinol

Precaución: for Research Use Only

Formula: C₅H₂BrF₃N₂O
MW: 242.99 g/mol
MDL Number: MFCD09901776
CAS Number: 942060-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR