5-Chloro-6-hydroxynicotinaldehyde 95%

Supplier: Ambeed
Warning

A689194-100MG A689194-1G A689194-250MG
77035-726EA 13 USD
77035-726 77035-728 77035-730
5-Chloro-6-hydroxynicotinaldehyde 95%
5-Chloro-6-hydroxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MDL Number: MFCD04972384
CAS Number: 627501-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR