Methyl-5-methylthiophene-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-methylthiophene-3-carboxylate
A687965-100MG
A687965-250MG
A687965-5G
77035-720EA
7
USD
77035-720
77035-722
77035-724
Methyl-5-methylthiophene-3-carboxylate 97%
Methyl-5-methylthiophene-3-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₈O₂S |
MDL Number:
MFCD02245126 CAS Number: 88770-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...