Methyl 3,4-bis(benzyloxy)benzoate 97%

Supplier: Ambeed
Warning

A687556-1G A687556-250MG
77018-626EA 45 USD
77018-626 77018-628
Methyl 3,4-bis(benzyloxy)benzoate 97%
Methyl 3,4-bis(benzyloxy)benzoate

Precaución: for Research Use Only

Formula: C₂₂H₂₀O₄
MDL Number: MFCD06203841
CAS Number: 54544-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR