Diethyl furan-2,5-dicarboxylate 97%

Supplier: Ambeed
Warning

A687533-10G A687533-250MG A687533-25G A687533-5G A687533-100G
77035-708EA 27 USD
77035-708 77035-710 77035-712 77035-714 77035-716 77873-086
Diethyl furan-2,5-dicarboxylate 97%
Diethyl furan-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₅
MDL Number: MFCD00449457
CAS Number: 53662-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR