α,3-Dibromo-2-fluorotoluene 97%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-3-(bromomethyl)-2-fluorobenzene, 3-Bromo-2-fluorobenzyl bromide

A687049-10G A687049-1G A687049-250MG A687049-25G A687049-5G
77005-912EA 121 USD
77005-912 77005-914 77005-916 77005-918 77005-920
α,3-Dibromo-2-fluorotoluene 97%
α,3-Dibromo-2-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MDL Number: MFCD09842442
CAS Number: 149947-16-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR