N,N-Dibenzyl-4-bromoaniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-N,N-dibenzylaniline

A686550-1G A686550-250MG
77035-700EA 72.15 USD
77035-700 77035-702
N,N-Dibenzyl-4-bromoaniline 95%
N,N-Dibenzyl-4-bromoaniline

Precaución: for Research Use Only

Formula: C₂₀H₁₈BrN
MW: 352.28 g/mol
MDL Number: MFCD09037473
CAS Number: 65145-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR