5-Bromo-2-(2,2,2-trifluoroethoxy)pyrimidine 95%

Supplier: Ambeed
Warning

A685182-1G A685182-250MG A685182-5G
77018-610EA 45 USD
77018-610 77018-612 77018-614
5-Bromo-2-(2,2,2-trifluoroethoxy)pyrimidine 95%
5-Bromo-2-(2,2,2-trifluoroethoxy)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂O
MDL Number: MFCD07375136
CAS Number: 433683-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR