Ethyl 1H-pyrazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A683365-1G A683365-5G
77035-694EA 11 USD
77035-694 77669-858
Ethyl 1H-pyrazole-5-carboxylate 95%
Ethyl 1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD00464260
CAS Number: 21056-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR