5-Chloro-2-thiophenesulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

A683360-10G A683360-1G A683360-5G A683360-25G
77005-894EA 21.22 USD
77005-894 77005-896 77005-898 77673-866
5-Chloro-2-thiophenesulfonyl chloride 97%
5-Chloro-2-thiophenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
MDL Number: MFCD00051667
CAS Number: 2766-74-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR