4-Nitrophenyl acetate 97%

Supplier: Ambeed
Warning

A683129-100G A683129-5G A683129-25G
76992-604EA 82 USD
76992-604 76992-606 77614-112
4-Nitrophenyl acetate 97%
4-Nitrophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00007326
CAS Number: 830-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR