4-Bromo-3-chloro-2-nitroaniline 95%

Supplier: Ambeed
Warning

A683083-100MG A683083-250MG
76992-600EA 37 USD
76992-600 76992-602
4-Bromo-3-chloro-2-nitroaniline 95%
4-Bromo-3-chloro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O₂
MDL Number: MFCD09878187
CAS Number: 1000573-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR