7-Bromo-2-ethoxyquinoline 95%

Supplier: Ambeed
Warning

A683052-1G A683052-250MG
77018-588EA 24 USD
77018-588 77018-590
7-Bromo-2-ethoxyquinoline 95%
7-Bromo-2-ethoxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO
MW: 252.10 g/mol
MDL Number: MFCD25956746
CAS Number: 1223559-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR