2-Naphthyl phenyl ketone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Benzoylnaphthalene

A682951-1G A682951-250MG A682951-25G A682951-5G
77035-686EA 39.26 USD
77035-686 77035-688 77035-690 77035-692
2-Naphthyl phenyl ketone 98%
2-Naphthyl phenyl ketone

Precaución: for Research Use Only

Formula: C₁₇H₁₂O
MW: 232.28 g/mol
MDL Number: MFCD00004106
CAS Number: 644-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR