5-Bromo-2-(dimethylamino)nicotinonitrile 95+%
Supplier: Ambeed
|
Warning
|
A682795-100MG
A682795-1G
A682795-250MG
76992-594EA
97.61
USD
76992-594
76992-596
76992-598
5-Bromo-2-(dimethylamino)nicotinonitrile 95+%
5-Bromo-2-(dimethylamino)nicotinonitrile
Precaución: for Research Use Only
Formula:
C₈H₈BrN₃ MW: 226.07 g/mol |
MDL Number:
MFCD19688659 CAS Number: 1346537-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...