2-Piperazin-1-yl-1,3-benzoxazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Piperazin-1-yl)benzo[d]oxazole

A682527-100MG A682527-250MG A682527-5G
77018-580EA 31.83 USD
77018-580 77018-582 77018-584
2-Piperazin-1-yl-1,3-benzoxazole 98%
2-Piperazin-1-yl-1,3-benzoxazole

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
MDL Number: MFCD04035522
CAS Number: 111628-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR