N3-Methylpyridine-2,3-diamine 97%

Supplier: Ambeed
Warning

A682040-1G A682040-5G A682040-100MG
76992-590EA 187.78 USD
76992-590 76992-592 77684-586
N3-Methylpyridine-2,3-diamine 97%
N3-Methylpyridine-2,3-diamine

Precaución: For research use only.

Formula: C₆H₉N₃
MW: 123.156 g/mol
MDL Number: MFCD00490112
CAS Number: 56291-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR