2,5-Dibromo-3-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A681575-100G
A681575-10G
A681575-1G
A681575-25G
A681575-500G
A681575-5G
77005-874EA
59
USD
77005-874
77005-876
77005-878
77005-880
77005-882
77005-884
2,5-Dibromo-3-nitropyridine 98%
2,5-Dibromo-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₅H₂Br₂N₂O₂ |
MDL Number:
MFCD09266223 CAS Number: 15862-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...