7-Bromo-5-methylbenzo[b]furan 98%

Supplier: Ambeed
Warning

A680686-1G A680686-250MG A680686-5G
77018-554EA 17 USD
77018-554 77018-556 77018-558
7-Bromo-5-methylbenzo[b]furan 98%
7-Bromo-5-methylbenzo[b]furan

Precaución: for Research Use Only

Formula: C₉H₇BrO
MDL Number: MFCD11877828
CAS Number: 35700-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR