2,2-Pentamethylene-1,3-dioxolane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,4-Dioxaspiro[4.5]decane
A680640-1G
A680640-5G
A680640-250MG
77018-552EA
46.68
USD
77018-552
77684-584
77684-583
2,2-Pentamethylene-1,3-dioxolane 97%
2,2-Pentamethylene-1,3-dioxolane
Precaución: For research use only.
Formula:
C₈H₁₄O₂ MW: 142.196 g/mol |
MDL Number:
MFCD00005409 CAS Number: 177-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...