(S)-4-Benzyl-3-(2-chloroacetyl)oxazolidin-2-one 98+%

Supplier: Ambeed
Warning

A680392-100MG A680392-250MG
77035-674EA 48.81 USD
77035-674 77035-676
(S)-4-Benzyl-3-(2-chloroacetyl)oxazolidin-2-one 98+%
(S)-4-Benzyl-3-(2-chloroacetyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClNO₃
MW: 253.68 g/mol
MDL Number: MFCD00269662
CAS Number: 104324-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR