Methyl(1-phenylethyl)amine 98%

Supplier: Ambeed
Danger

A680270-1G A680270-25G A680270-5G
77035-664EA 29 USD
77035-664 77035-666 77573-202
Methyl(1-phenylethyl)amine 98%
Methyl(1-phenylethyl)amine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MDL Number: MFCD00463429
CAS Number: 32512-24-6
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR