Ethyl 8-(benzyloxy)imidazo[1,2-a]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A680183-100MG A680183-1G A680183-250MG
77018-546EA 41.38 USD
77018-546 77018-548 77018-550
Ethyl 8-(benzyloxy)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 8-(benzyloxy)imidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₃
MW: 296.32 g/mol
MDL Number: MFCD06739214
CAS Number: 79707-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR