2,2',2''-(2,4,6-Trifluorobenzene-1,3,5-triyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 98%

Supplier: Ambeed
Warning

A679637-100MG A679637-1G A679637-250MG A679637-5G
77018-538EA 39 USD
77018-538 77018-540 77018-542 77018-544
2,2',2''-(2,4,6-Trifluorobenzene-1,3,5-triyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 98%
2,2',2''-(2,4,6-Trifluorobenzene-1,3,5-triyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Precaución: for Research Use Only

Formula: C₂₄H₃₆B₃F₃O₆
MDL Number: MFCD12407207
CAS Number: 408492-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR