2-(4-Methoxy-2-nitrophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A679605-100MG A679605-1G A679605-250MG A679605-5G
77018-530EA 26.53 USD
77018-530 77018-532 77018-534 77018-536
2-(4-Methoxy-2-nitrophenyl)acetic acid 95%
2-(4-Methoxy-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MW: 211.17 g/mol
MDL Number: MFCD06659779
CAS Number: 20876-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR